Assay Details
| Gene Symbol: GOSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012527.3 | XP_004012576.1 | 1 - 2 | 107 | 79 | |
| XM_012185431.2 | XP_012040821.1 | 1 - 2 | 108 | 79 | ||
More Information
Set Membership: |
|

GOSR1
Genomic Map