Assay Details
Gene Symbol: HEXD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013048.4 | XP_004013097.3 | 7 - 8 | 829 | 87 | |
XM_027974312.1 | XP_027830113.1 | 7 - 8 | 829 | 87 | ||
XM_027974313.1 | XP_027830114.1 | 7 - 8 | 829 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
