Assay Details
| Gene Symbol: RIMKLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015094957.2 | XP_014950443.1 | 5 - 6 | 825 | 65 | |
| XM_027967899.1 | XP_027823700.1 | 5 - 6 | 883 | 65 | ||
| XM_027967900.1 | XP_027823701.1 | 5 - 6 | 787 | 65 | ||
More Information
Set Membership: |
|

RIMKLB
Genomic Map