Assay Details
Gene Symbol: LOC101113487 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004007023.3 | XP_004007072.1 | 7 - 8 | 882 | 67 | |
XM_012175714.2 | XP_012031104.1 | 7 - 8 | 882 | 67 | ||
XM_012175715.3 | XP_012031105.1 | 7 - 8 | 875 | 67 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
