Assay Details
| Gene Symbol: DTX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967525.1 | XP_027823326.1 | 4 - 5 | 971 | 72 | |
| XM_027967526.1 | XP_027823327.1 | 4 - 5 | 1311 | 72 | ||
More Information
Set Membership: |
|

DTX3
Genomic Map