Assay Details
Gene Symbol: LOC101120058 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004005253.4 | XP_004005302.3 | 15 - 16 | 1803 | 71 | |
XM_027964322.1 | XP_027820123.1 | 14 - 15 | 1731 | 71 |
Gene Symbol: LOC114112956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964321.1 | XP_027820122.1 | 4 - 5 | 438 | 71 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
