Assay Details
| Gene Symbol: LOC101117429 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963808.1 | XP_027819609.1 | 6 - 7 | 758 | 93 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101117429| Gene Symbol: LOC101117429 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963808.1 | XP_027819609.1 | 6 - 7 | 758 | 93 | |