Assay Details
| Gene Symbol: GFRAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015102641.2 | XP_014958127.2 | 5 - 6 | 699 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GFRAL| Gene Symbol: GFRAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015102641.2 | XP_014958127.2 | 5 - 6 | 699 | 65 | |
Set Membership: |
|