Assay Details
Gene Symbol: SHLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962427.1 | XP_027818228.1 | 5 - 6 | 2010 | 79 | |
XM_027962428.1 | XP_027818229.1 | 5 - 6 | 2022 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
