Assay Details
Gene Symbol: THAP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979555.1 | XP_027835356.1 | 2 - 3 | 327 | 98 | |
XM_027979557.1 | XP_027835358.1 | 2 - 3 | 327 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
