Assay Details
| Gene Symbol: TRMT10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015096398.2 | XP_014951884.2 | 4 - 5 | 632 | 78 | |
| XM_015096399.2 | XP_014951885.2 | 4 - 5 | 978 | 78 | ||
More Information
Set Membership: |
|

TRMT10A
Genomic Map