Assay Details
Gene Symbol: PRR11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012405.4 | XP_004012454.2 | 9 - 10 | 1114 | 60 | |
XM_027975345.1 | XP_027831146.1 | 9 - 10 | 1122 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
