Assay Details
| Gene Symbol: MTREX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016983.4 | XP_004017032.1 | 23 - 24 | 2770 | 72 | |
| XM_027980070.1 | XP_027835871.1 | 19 - 20 | 2308 | 72 | ||
More Information
Set Membership: |
|

MTREX
Genomic Map