Assay Details
Gene Symbol: LOC101120580 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971620.1 | XP_027827421.1 | 22 - 23 | 3095 | 110 | |
XM_027971621.1 | XP_027827422.1 | 23 - 24 | 3361 | 110 |

More Information
Set Membership: |
![]() ![]() |
