Assay Details
Gene Symbol: LOC114112296 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963775.1 | XP_027819576.1 | 1 - 2 | 141 | 64 |
Gene Symbol: LOC101115632 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973653.1 | XP_027829454.1 | 5 - 6 | 433 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027963775.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
