Assay Details
Gene Symbol: SCN2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004640.4 | XP_004004689.2 | 11 - 12 | 1919 | 66 | |
XM_004004641.4 | XP_004004690.2 | 11 - 12 | 2837 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
