Assay Details
| Gene Symbol: PCBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957565.1 | XP_027813366.1 | 8 - 9 | 846 | 73 | |
| XM_027957566.1 | XP_027813367.1 | 9 - 10 | 833 | 73 | ||
| XM_027957567.1 | XP_027813368.1 | 9 - 10 | 921 | 73 | ||
| XM_027957568.1 | XP_027813369.1 | 8 - 9 | 744 | 73 | ||
| XM_027957570.1 | XP_027813371.1 | 9 - 10 | 833 | 73 | ||
| XM_027957571.1 | XP_027813372.1 | 9 - 10 | 830 | 73 | ||
| XM_027957572.1 | XP_027813373.1 | 8 - 9 | 847 | 73 | ||
More Information
Set Membership: |
|

PCBP4
Genomic Map