Assay Details
Gene Symbol: LOC101116707 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964869.1 | XP_027820670.1 | 11 - 12 | 1097 | 118 | |
XM_027964870.1 | XP_027820671.1 | 11 - 12 | 1129 | 118 | ||
XM_027964871.1 | XP_027820672.1 | 10 - 11 | 1001 | 118 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
