Assay Details
Gene Symbol: LOC101116707 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964869.1 | XP_027820670.1 | 13 - 14 | 1268 | 78 | |
XM_027964870.1 | XP_027820671.1 | 13 - 14 | 1300 | 78 | ||
XM_027964871.1 | XP_027820672.1 | 12 - 13 | 1172 | 78 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
