Assay Details
Gene Symbol: DNTTIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012176567.3 | XP_012031957.2 | 1 - 2 | 127 | 64 | |
XM_027972219.1 | XP_027828020.1 | 1 - 2 | 128 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
