Assay Details
Gene Symbol: LOC101117600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013284.4 | XP_004013333.1 | 5 - 6 | 649 | 68 | |
XM_027975008.1 | XP_027830809.1 | 4 - 5 | 554 | 68 | ||
XM_027975009.1 | XP_027830810.1 | 4 - 5 | 535 | 68 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
