Assay Details
Gene Symbol: TXNIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002420.4 | XP_004002469.1 | 6 - 7 | 1268 | 72 | |
GenBank mRNA | DQ314291.1 | - | 2 - 3 | 252 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
