Assay Details
Gene Symbol: PTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008093.3 | XP_004008142.2 | 3 - 4 | 735 | 89 | |
XM_027968848.1 | XP_027824649.1 | 3 - 4 | 735 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
