Assay Details
| Gene Symbol: LOC101118895 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015970.4 | XP_004016019.2 | 1 - 2 | 102 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101118895| Gene Symbol: LOC101118895 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015970.4 | XP_004016019.2 | 1 - 2 | 102 | 76 | |
Set Membership: |
|