Assay Details
Gene Symbol: LOC101120466 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964020.1 | XP_027819821.1 | 16 - 17 | 2113 | 69 |
Gene Symbol: LOC114112915 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964009.1 | XP_027819810.1 | 2 - 3 | 295 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
