Assay Details
| Gene Symbol: LOC101115486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012098423.3 | XP_011953813.2 | 14 - 15 | 4801 | 69 | |
| XM_012098428.3 | XP_011953818.2 | 9 - 10 | 2785 | 69 | ||
| XM_027975368.1 | XP_027831169.1 | 18 - 19 | 5508 | 69 | ||
| XM_027975369.1 | XP_027831170.1 | 18 - 19 | 5508 | 69 | ||
| XM_027975370.1 | XP_027831171.1 | 18 - 19 | 5508 | 69 | ||
| XM_027975371.1 | XP_027831172.1 | 18 - 19 | 5508 | 69 | ||
| XM_027975372.1 | XP_027831173.1 | 17 - 18 | 5442 | 69 | ||
| XM_027975373.1 | XP_027831174.1 | 16 - 17 | 5388 | 69 | ||

LOC101115486
Genomic Map