Assay Details
Gene Symbol: CNNM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006156.4 | XP_004006205.3 | 6 - 7 | 2138 | 62 | |
XM_027967093.1 | XP_027822894.1 | 7 - 8 | 2201 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
