Assay Details
| Gene Symbol: GRIN2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978558.1 | XP_027834359.1 | 2 - 3 | 1083 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GRIN2D| Gene Symbol: GRIN2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978558.1 | XP_027834359.1 | 2 - 3 | 1083 | 66 | |
Set Membership: |
|