Assay Details
| Gene Symbol: CCDC115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004476.4 | XP_004004525.1 | 5 - 6 | 704 | 71 | |
| XM_012130041.3 | XP_011985431.1 | 4 - 5 | 963 | 71 | ||
| XM_027964654.1 | XP_027820455.1 | 5 - 6 | 730 | 71 | ||
| XM_027964655.1 | XP_027820456.1 | 5 - 6 | 673 | 71 | ||
| XM_027964656.1 | XP_027820457.1 | 5 - 6 | 658 | 71 | ||
| XM_027964657.1 | XP_027820458.1 | 5 - 6 | 650 | 71 | ||
| XM_027964658.1 | XP_027820459.1 | 4 - 5 | 414 | 71 | ||
More Information
Set Membership: |
|

CCDC115
Genomic Map