Assay Details
| Gene Symbol: TMEM59L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969389.1 | XP_027825190.1 | 2 - 3 | 404 | 69 | |
| XM_027969390.1 | XP_027825191.1 | 2 - 3 | 404 | 69 | ||
More Information
Set Membership: |
|

TMEM59L
Genomic Map