Assay Details
| Gene Symbol: LOC101105495 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015096090.2 | XP_014951576.2 | 1 - 2 | 3641 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101105495| Gene Symbol: LOC101105495 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015096090.2 | XP_014951576.2 | 1 - 2 | 3641 | 73 | |
Set Membership: |
|