Assay Details
Gene Symbol: LOC101123372 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012177962.3 | XP_012033352.1 | 5 - 6 | 479 | 75 | |
XM_012177964.3 | XP_012033354.1 | 5 - 6 | 479 | 75 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
