Assay Details
Gene Symbol: LOC101114198 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011124.4 | XP_004011173.2 | 2 - 3 | 878 | 118 | |
XM_012182449.3 | XP_012037839.2 | 2 - 3 | 878 | 118 | ||
XM_012182451.3 | XP_012037841.2 | 2 - 3 | 879 | 118 |

More Information
Set Membership: |
![]() ![]() |
