Assay Details
Gene Symbol: LOC101114198 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011124.4 | XP_004011173.2 | 24 - 25 | 3990 | 82 | |
XM_012182449.3 | XP_012037839.2 | 24 - 25 | 3993 | 82 | ||
XM_012182451.3 | XP_012037841.2 | 24 - 25 | 3994 | 82 |
Gene Symbol: LOC114116228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972738.1 | XP_027828539.1 | 2 - 3 | 283 | 82 |
