Assay Details
Gene Symbol: LOC101114198 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011124.4 | XP_004011173.2 | 7 - 8 | 1494 | 89 | |
XM_012182449.3 | XP_012037839.2 | 7 - 8 | 1494 | 89 | ||
XM_012182451.3 | XP_012037841.2 | 7 - 8 | 1495 | 89 |
