Assay Details
| Gene Symbol: RPS19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015294.3 | XP_004015343.1 | 5 - 6 | 882 | 99 | |
| XM_027978401.1 | XP_027834202.1 | 5 - 6 | 429 | 99 | ||
More Information
Set Membership: |
|

RPS19
Genomic Map