Assay Details
Gene Symbol: TMEM267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017009.4 | XP_004017058.2 | 3 - 4 | 1287 | 66 | |
XM_004017010.3 | XP_004017059.2 | 3 - 4 | 626 | 66 | ||
XM_012096966.2 | XP_011952356.2 | 4 - 5 | 792 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
