Assay Details
Gene Symbol: PTER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014243.4 | XP_004014292.2 | 4 - 5 | 999 | 70 | |
XM_012188435.3 | XP_012043825.2 | 5 - 6 | 1090 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
