Assay Details
| Gene Symbol: LOC101118459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098780.2 | XP_014954266.2 | 5 - 6 | 1138 | 77 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101118459| Gene Symbol: LOC101118459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098780.2 | XP_014954266.2 | 5 - 6 | 1138 | 77 | |