Assay Details
Gene Symbol: ELOVL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016954.4 | XP_004017003.2 | 2 - 3 | 184 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
