Assay Details
| Gene Symbol: GMPPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012100053.3 | XP_011955443.1 | 8 - 9 | 907 | 57 | |
| XM_012100055.3 | XP_011955445.1 | 7 - 8 | 1012 | 57 | ||
| XM_027957632.1 | XP_027813433.1 | 8 - 9 | 904 | 57 | ||
More Information
Set Membership: |
|

GMPPB
Genomic Map