Assay Details
Gene Symbol: SAPCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004018939.4 | XP_004018988.2 | 1 - 2 | 390 | 70 | |
XM_015102788.2 | XP_014958274.2 | 1 - 2 | 390 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
