Assay Details
| Gene Symbol: RAP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012178534.3 | XP_012033924.1 | 7 - 8 | 622 | 74 | |
| XM_027973069.1 | XP_027828870.1 | 7 - 8 | 614 | 74 | ||
| XM_027973074.1 | XP_027828875.1 | 6 - 7 | 536 | 74 | ||
| XM_027973076.1 | XP_027828877.1 | 6 - 7 | 486 | 74 | ||
| XM_027973081.1 | XP_027828882.1 | 5 - 6 | 517 | 74 | ||
More Information
Set Membership: |
|

RAP1A
Genomic Map