Assay Details
| Gene Symbol: C19H3orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015102430.2 | XP_014957916.2 | 2 - 3 | 1905 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C19H3orf62| Gene Symbol: C19H3orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015102430.2 | XP_014957916.2 | 2 - 3 | 1905 | 107 | |
Set Membership: |
|