Assay Details
| Gene Symbol: PHACTR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012183029.2 | XP_012038419.1 | 8 - 9 | 1736 | 56 | |
| XM_015097454.2 | XP_014952940.1 | 8 - 9 | 1558 | 56 | ||
| XM_027972510.1 | XP_027828311.1 | 8 - 9 | 1736 | 56 | ||
| XM_027972511.1 | XP_027828312.1 | 7 - 8 | 1534 | 56 | ||
| XM_027972512.1 | XP_027828313.1 | 7 - 8 | 1534 | 56 | ||
| XM_027972513.1 | XP_027828314.1 | 6 - 7 | 1337 | 56 | ||
More Information
Set Membership: |
|

PHACTR2
Genomic Map