Assay Details
Gene Symbol: TTLL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975828.1 | XP_027831629.1 | 6 - 7 | 5431 | 59 | |
XM_027975829.1 | XP_027831630.1 | 6 - 7 | 5431 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
