Assay Details
Gene Symbol: HERPUD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015015.3 | XP_004015064.1 | 5 - 6 | 665 | 60 | |
XM_027977858.1 | XP_027833659.1 | 5 - 6 | 640 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
