Assay Details
| Gene Symbol: LOC101118184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979067.1 | XP_027834868.1 | 3 - 4 | 418 | 89 | |
| XM_027979069.1 | XP_027834870.1 | 3 - 4 | 440 | 89 | ||
More Information
Set Membership: |
|

LOC101118184
Genomic Map