Assay Details
Gene Symbol: LOC101102057 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008561.4 | XP_004008610.1 | 5 - 6 | 445 | 97 | |
XM_027969726.1 | XP_027825527.1 | 4 - 5 | 355 | 97 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
