Assay Details
Gene Symbol: MAPK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012181550.3 | XP_012036940.1 | 2 - 3 | 1283 | 86 | |
XM_027971825.1 | XP_027827626.1 | 2 - 3 | 1333 | 86 | ||
XM_027971827.1 | XP_027827628.1 | 2 - 3 | 1347 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
